Upright and chest laboratory ultra-freezers, PRATILAB, IRILAB
This ultra freezer line offers a wide range of upright and chest models, with smart division of the interior space for storing samples, sophisticated control devices (also PC linked) and a large range of accessories (available on request). Units have 140 mm high pressure (40 kg/cm) polyurethane foam insulation.
- Sample safety: the microprocessor Setting and Control system has two independent sections: temperature control, alarms managements and temperature display
- Ethernet Interface provides a connection to an external PC - to monitor alarms and parameters status
- Buffer battery maintains the system active for 24 hours
- Electromagnetic opening system with standard key
- Rollers and adjustable feet for easy positioning
Temperature range: −40 to −85 °C
- Catalog No:
- 471-0891
- 471-0899
- 471-0892
- 471-0890
- 471-0889
- 471-0888
- Model:
- PLATILAB 550
- IRILAB 800
- PLATILAB 110
- PLATILAB 370
- PRATILAB 500
- PRATILAB 340
- Cat. No.:
- 471-0891
- 471-0899
- 471-0892
- 471-0890
- 471-0889
- 471-0888
- Display:
- Graphic
- Graphic
- Graphic
- Graphic
- Graphic
- Graphic
- Alarm:
- Audible and visual
- Audible and visual
- Audible and visual
- Audible and visual
- Audible and visual
- Audible and visual
- Shelves/drawers:
- 3 adjustable stainless steel shelves
- 3 adjustable stainless steel shelves
- 3 adjustable stainless steel shelves
- Door type:
- 1 outer, 1 inner door with special magnetic gaskets
- 1 outer, 1 inner door
- 1 outer, 1 inner door with special magnetic gaskets
- 1 outer, 1 inner door with special magnetic gaskets
- 1 outer, 1 inner door with special magnetic gaskets
- 1 outer, 1 inner door with special magnetic gaskets
- Door lock:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Access ports:
- standard port hole (Ø 23 mm)
- standard port hole (Ø 23 mm)
- standard port hole (Ø 23 mm)
- standard port hole (Ø 23 mm)
- standard port hole (Ø 23 mm)
- standard port hole (Ø 23 mm)
Frequently Bought Together