Burettes, lateral stopcock, class B, amber glass
Supplier: HIRSCHMANN
Certificates
About this item
Burette, class B, DURAN®, brown glass.
- White graduation marks
- Lateral stopcock with glass key
- Brown coloured for light sensitive substances
Certifications: Calibrated for EX, DIN EN ISO 385.
Frequently Bought Together