Burettes, lateral stopcock, Schellbach
Supplier: HIRSCHMANN
Certificates
About this item
Burette, class B, DURAN® borosilicate glass 3.3, with blue graduation.
- Lateral glass stopcock with glass key or PTFE spindle
- Schellbach stripes for enhanced legibility
Certifications: Calibrated for EX, DIN EN ISO 385.
Frequently Bought Together