Order Entry
Hungary
ContactUsLinkComponent
Pressure filtration holders, stainless steel
Pressure filtration holders, stainless steel
Supplier:  Sartorius
Certificates
undefined
Pressure filtration holders, stainless steel
Supplier:  Sartorius
Specifications
  • Catalog No:
  • SART16251
  • SART16278
  • 512-3810
  • Filtration area:
  • 3 cm²
  • 13 cm²
  • 13 cm²
  • Max. operating pressure:
  • 5 bar
  • 5 bar
  • 5 bar
  • Weight:
  • ∼ 170 g
  • ∼ 490 g