Order Entry
Hungary
ContactUsLinkComponent
Pipette tray
Pipette tray
  BRND28300
 :  Brand
undefined
Pipette tray
  BRND28300
 :  Brand
  • Description:
    Pipette tray, white PVC
  • W×D×H:
    280×220×39 mm
  • Pk:
    1
Made of white PVC.
  • Good temperature resistance
  • Good chemical resistance
  • Easy to clean
Practical tray for pipettes of any size.