4-Ethylbenzenesulphonyl chloride
Catalog # MOLEM52111404
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:204.68 g/mol
Boiling Pt:143 °C (16 mmHg)
Melting Pt:12 °C
Density:1.268 g/cm³ (20 °C)
UN:3265
ADR:8,II
CAS Number:16712-69-9
Formula:C₈H₉ClO₂S
MDL Number:MFCD00041480