Order Entry
Hungary
ContactUsLinkComponent
 
Sodium thiosulphate 0.01 mol/l (0.01 N) in aqueous solution
Catalog # ROTHX869.1
Supplier:  Roth Carl
CAS Number:  
Sodium thiosulphate 0.01 mol/l (0.01 N) in aqueous solution
Catalog # ROTHX869.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

About this item

 :
 :158.11 g/mol
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499