Order Entry
Hungary
Orders LinkContactUsLinkComponent
VWR® Traceable® Infrared Thermometer

VWR® Traceable® Infrared Thermometer

−55...+300
(−55...+250 °C*/−55…+330 °C**), ±0,6; ±0,5* (5…65) °C, 0,2 (−10…+200); 1,0 °C, 165×38×2,5 mm, 100 g
Catalog # 620-1806
Supplier:   VWR Collection
VWR® Traceable® Infrared Thermometer
Catalog # 620-1806
Supplier:  VWR Collection
VWR® Traceable® Infrared Thermometer
VWR® Traceable® Infrared Thermometer

−55...+300
(−55...+250 °C*/−55…+330 °C**), ±0,6; ±0,5* (5…65) °C, 0,2 (−10…+200); 1,0 °C, 165×38×2,5 mm, 100 g

Price
117 000 Ft
Log in for availability
1 * 1 DB
Product Details & Documents

This HACCP - Hazard Analysis and Critical Control Point - thermometer is designed for food laboratories, food processing, food preparation and brewing. The waterproof construction allows it to be used in labs, plants and wash-down areas.

Infrared* or foldable stainless steel probe** (152 mm).

W×D×H: 165×38×2,5 mm

Weight: 100 g

Delivery information: Supplied with serial numbered NIST Traceable® certificate of calibration.

Specifications
  • Range:
    −55...+300
    (−55...+250 °C*/−55…+330 °C**)
  • Resolution:
    0,2 (−10…+200); 1,0 °C
  • Weight:
    100 g
  • W×D×H:
    165×38×2,5 mm
  • Accuracy:
    ±0,6; ±0,5* (5…65) °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.