Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Melting Point Apparatus
Accessories for melting point apparatus
Accessories for melting point apparatus
Accessories for melting point apparatus
621-0016
:
Cole-Parmer
Accessories for melting point apparatus
621-0016
:
Cole-Parmer
Description:
AZ6730 calibration kit
Seq:
11
Pk:
1