Test tubes without rim, heavy wall
Supplier: MURRAY S
Certificates
About this item
Borosilicate glass, grade 3.3, SAMCO '33'. Thick walled.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Suitable for all usual laboratory applications. They will withstand temperature change up to and including boiling of samples.
Certifications: ISO 4142:2002
Frequently Bought Together