Sterilising pans, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
PP.
- Heavy duty construction
- Extra strong rims
For repeated autoclaving of instruments and labware.
Specifications
- Catalog No:
- 489-2142
- 489-2143
- 489-2140
- 489-2141
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement