Instrument trays
Supplier: P.S. KUNSTSTOFWERKE
Certificates
About this item
Melamine, white, polished.
- Good chemical resistance
- Heat resistant up to 80 °C
- Robust, smooth surfaces for easy cleaning
Low profile.
Frequently Bought Together
Melamine, white, polished.
Low profile.