Chemical spoon with spatula end
Supplier: BOCHEM
Certificates
About this item
Stainless steel 18/10, anti-magnetic or stainless steel with PTFE coating. Multipurpose spoon for a wide range of uses.
- With flat handle
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Quick and easy to clean
Frequently Bought Together