Order Entry
Hungary
ContactUsLinkComponent
 
Deflagrating spoon
  231-1272
undefined
Deflagrating spoon
  231-1272

 

  • Length:
    450 mm
  • Pk:
    1
  • Spoon size:
    16 mm

 

 

Made of stainless steel, cup Ø 16 mm. Total length: 450 mm.