Test tube racks
: Brand
PP, coloured.
- Filled PP, density 1,2 g/cm³ so will not float in waterbath
- Operating temperature −20 to 90 °C
- Autoclavable (121 °C), (2 bar), acc. DIN EN 285
Versatile and inexpensive racks for common diameter test tubes. Alphanumerical positions for clear sample identification. Wide spaces between positions. The racks are delivered dismantled and can be put together securely and permanently in a few steps, without tools.
Frequently Bought Together