VWR®, Vacuum/Pressure Bulb
Catalog # 612-2705
Supplier: VWR Collection
Specifications
- Description:Vacuum/pressure bulb without tubing
- Pk:1
Specifications
About this item
Black rubber with PVC connectors.
- For collecting gas-phase samples or aspirating gases through instruments
- Two thick-walled rubber valves
Capacity: 85 ml
Colour: black
Ordering information: Supplied without tubing.