Vacuum filtration units
Supplier: Sartorius
Certificates
About this item
Borosilicate glass, anodised aluminium clamp, silicone lid, autoclavable.
Compact holder for easy particle testing and colony counting, clarification and sterile filtration.
516-7016: With glass frit filter support
512-3321: With PTFE-coated stainless steel screen filter support
Frequently Bought Together