Tweezers
: VITLAB
POM, yellow. Universal tweezers for a variety of applications.
- Blunt, elastic, very good resilience
- With grooves on the outside for optimum handling and grip
- Good chemical resistance
Frequently Bought Together
POM, yellow. Universal tweezers for a variety of applications.