VWR®, Dishes, Crystallising
Borosilicate 3.3 glass, with or without spout.
- Very good chemical resistance
- High temperature resistance up to +500 °C
- Minimal thermal expansion, giving relatively high resistance to temperature changes
The economical alternative for a wide range of applications. With white labelling area.
DIN 12337 (without spout)
DIN 12338 (with spout)
Frequently Bought Together