Desiccator plates
Supplier: MORGAN ADVANCED MATERIALS HALD
Certificates
About this item
Porcelain desiccator plates. Central hole 20 mm Ø, filter holes ~5 mm Ø. Suitable for use with any desiccators with corresponding diameter.
Certifications: Conform to DIN 12911
Frequently Bought Together