Tubing clamps, screw style, SP Bel-Art
Catalog # 229-0007
Supplier: Bel-Art Products, a Part of SP
Specifications
- Spanning width:14 mm
- Colour:White
- Pk:3
Specifications
About this item
Molded of Acetal plastic. This tubing clamp offers on/off and fine control so accurate that it replaces stopcocks in many applications.
- Can be installed without disconnecting tubing or interrupting operation
- Accepts flexible tubing up to 14 mm Ext.Ø
- Resistant to acids, alkalis and organic solvents
- Autoclavable