Order Entry
Hungary
ContactUsLinkComponent
VWR®, Clamps, Three Pronged
VWR®, Clamps, Three Pronged
Catalog # 241-0015
Supplier:  VWR Collection
undefined
VWR®, Clamps, Three Pronged
Catalog # 241-0015
Supplier:  VWR Collection
Specifications
  • Spanning width:
    0 - 30 mm
  • Pk:
    5
Specifications
About this item
Laboral aluminium alloy.
  • Clamping fingers covered in vinyl to protect clamped items
  • Easy action knurled screw for adjusting the clamping prongs