Burettes, straight stopcock, SILBERBRAND
Supplier: Brand
Certificates
About this item
Amber borosilicate glass 5.4.
- Calibrated to deliver 'TD, Ex'
- Short marks at major graduations, white graduations
25 ml: Tolerances according to class B, DIN EN ISO 385, reduced distance between subdivision marks.
50 ml: Class B, DIN EN ISO 385.
Length: 550 mm (25 ml); 800 mm (50 ml).
Frequently Bought Together