Burettes, straight stopcock, Schellbach
Supplier: HIRSCHMANN
Certificates
About this item
DURAN®, class B.
- Calibrated to deliver 'Ex'
- Short marks at major graduations, blue graduations
Scale length: 500 mm.
Scale length: 500 mm
Certifications: DIN ISO EN 385
Frequently Bought Together