Order Entry
Hungary
ContactUsLinkComponent
Brush for butyrometer
Brush for butyrometer
Catalog # HECH91120000012
undefined
Brush for butyrometer
Catalog # HECH91120000012
Specifications
  • Bristle material:
    Natural bristle
  • Handle material:
    Wire
  • Total length:
    260 mm
  • Pk:
    10
Specifications
About this item
Brush with natural bristles and two brush parts, outer 22×12 mm, inner 60×45 mm. Galvanized iron thread handle.