Waste disposal bags, Labsak, Jencons
129-1900
- Description:Bags, Labsak
- L×W:495×910 mm
- Material:PP
- Pk:25
Laminated polypropylene bag for general purpose laboratory and workshop use. Ideal for disposal purposes.
- Puncture and tear resistant
- Long term resistance to heat and UV and stabilised for weathering