Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 mol/l in aqueous solution
Supplier: SPEKTRUM-3D
Certificates
About this item
CAS Number:14634-91-4
:
Formula:C₃₆H₂₄FeN₆SO₄
MDL Number:MFCD00036428
:692.54 g/mol
Density:1.01 g/cm³ (20 °C)
EINECS:238-676-6
Frequently Bought Together