Order Entry
Hungary
ContactUsLinkComponent
BD® Arterial Cannula
BD® Arterial Cannula
Catalog # BDAM682245
Supplier:  BD Medical
BD® Arterial Cannula
Catalog # BDAM682245
Supplier:  BD Medical
Specifications
  • Flow rate:
    49 ml/min
  • Gauge:
    G 20
  • Sterile:
    +
  • Description:
    BD® Arterial cannula
  • Length:
    45 mm
  • Pk:
    25
Specifications
About this item
Designed specifically for arterial cannulation, the BD® arterial cannula enables arterial pressure monitoring and serial arterial blood gas determinations.
  • Developed to support smooth skin and vein penetration
  • Clear visibility of blood flashback confirms arterial entry
  • Flexible wings stabilise the catheter in place, helping to prevent unwanted movement
Device is designed to prevent back-flow, which potentially reduces the risk of air emboli and exposure to blood.