Amplite® Fluorimetric Lipase Activity Assay Kit
Catalog # AATB11322
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Pk:100 Tests
- Tests per kit:100 tests
Specifications
About this item
Designed for consistent and dependable results. The Amplite® Fluorimetric Lipase Activity Assay Kit provides a simple and quick protocol for measuring lipase activity.
- This kit is suitable for detecting lipases from various biological samples like cells, supernatants, tissue extracts, or serum, and is adaptable for high-throughput screening applications
- Ex/Em=540/610 nm
Lipases belong to a class of enzymes that catalyse the hydrolysis of ester bonds in lipids, such as triglycerides, phospholipids and cholesterol esters.
Delivery: Shipped with blue ice.
Caution: Research use only.