Anti-CD22 Mouse Recombinant Antibody [clone: rBLCAM/4108]
Catalog # ANTIA253793-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:B cell receptor CD22 precursor
- Antigen symbol:CD22
- Clonality:Recombinant
- Clone:rBLCAM/4108
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P20273
- Antigen synonyms:B lymphocyte cell adhesion molecule|CD 22|B-lymphocyte cell adhesion molecule|CD22 molecule|B-cell receptor CD22|BL-CAM|CD22 antigen|BL CAM|BLCAM
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:130 - 140 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 52-178, of human CD22 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rBLCAM/4108] antibody to CD22 for IHC-P with samples derived from human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD22
Clonality: Recombinant
Clone: rBLCAM/4108
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human