Anti-PKC alpha Rabbit Polyclonal Antibody
ANTIA11038-100
New Product
- Antibody type:Primary
- Antigen name:AAG6
- Antigen symbol:PKC alpha
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P17252
- Antigen synonyms:Aging associated gene 6|PKC-A|aPKC|KPCA_HUMAN|PRKACA|PKC-alpha|PRKCA|PKC alpha|PKCA
- Modification:phospho Ser319
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
- Molecular weight:70 kDa
- Sequence:VISPS
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthetic phosphorylated peptide around S319 of human PKC alpha (NP_002728.2)
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Rabbit polyclonal antibody to PKC alpha (phospho Ser319) for WB with samples derived from human, mouse and rat.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:1000
Type: Primary
Antigen: PKC alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat