Order Entry
Hungary
ContactUsLinkComponent
Anti-NCAM1 Rabbit Recombinant Antibody [clone: NCAM1/8392R]
Anti-NCAM1 Rabbit Recombinant Antibody [clone: NCAM1/8392R]
  ANTIA316759-100
New Product
Anti-NCAM1 Rabbit Recombinant Antibody [clone: NCAM1/8392R]
  ANTIA316759-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    antigen MSK39 identified by monoclonal
  • Antigen symbol:
    NCAM1
  • Clonality:
    Recombinant
  • Clone:
    NCAM1/8392R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P13591, P13592
  • Antigen synonyms:
    NCAM 1|NCAM|N-CAM-1|MSK39|CD56|antigen recognized by monoclonal|cell adhesion molecule, neural, 1|MSK 39|NCAM C
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
  • Molecular weight:
    180 kDa/145 kDa/125 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Recombinant fragment (around Amino Acids 600-800) of human NCAM1 (CD56) protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant rabbit monoclonal [NCAM1/8392R] antibody to NCAM1 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: NCAM1
Clonality: Recombinant
Clone: NCAM1/8392R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human