Human Recombinant TGF-beta1 (from CHO cells)
Catalog # ENZORLT-300-023
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Endotoxin content:<0,1 ng/mg of protein (<1 EU/mg)
- UniProtKB:P01137
- Protein/peptide name:TGF-beta1
- Purity:>98% by SDS-PAGE and HPLC analyses
- Molecular weight:25 kDa
- Formulation:Lyophilised
Specifications
About this item
Caution: Research use only.