Zinc(II) nitrate hexahydrate
MOLA3030201000
:
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
:
:297.49 g/mol
:12,10273
:∼105 °C (1013 hPa)
:36 °C
:2.06 g/cm³ (20 °C)
:Ambient
:231-943-8
:1514
:5.1,II
:GHS03
:GHS07
:10196-18-6
:ZnN₂O₆·6H₂O
:MFCD00149889