Sodium acetate trihydrate
MOLA3140221000
:
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
:
:136.08 g/mol
:13,08642
:120…123 °C (1013 hPa)
:58 °C
:1.4 g/cm³ (20 °C)
:Ambient
:204-823-8
:6131-90-4
:H₃CCOONa·3H₂O
:MFCD00071557