Bromothymol blue sodium salt
Catalog # MOLE24025529-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Melting Pt:> 200 °C
:646.37 g/mol
Storage Temperature:Ambient
EINECS:252-169-7
CAS Number:34722-90-2
Formula:C₂₇H₂₇Br₂NaO₅S
MDL Number:MFCD00077263