Order Entry
Hungary
ContactUsLinkComponent
DAF-2 diacetate 5 mM in DMSO ≥98% (by HPLC)
Irritant
DAF-2 diacetate 5 mM in DMSO ≥98% (by HPLC)
Catalog # ENZOALX620056M001
Supplier:  Enzo Life Sciences
CAS Number:  
Irritant
DAF-2 diacetate 5 mM in DMSO ≥98% (by HPLC)
Catalog # ENZOALX620056M001
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

Specification Test Results

  • Purity:
    ≥98% (HPLC)
  • Formulation:
    Dissolved in 0.45 ml DMSO.

Specification Test Results

About this item

 :
 :446.41 g/mol
EINECS:200-664-3
 :GHS07
Storage Temperature:Freezer
CAS Number:205391-02-2
Formula:C₂₄H₁₈N₂O₇
MDL Number:MFCD05865101