Order Entry
Hungary
ContactUsLinkComponent
Lipopolysaccharides (from S. abortus equi) S-form, TLRgrade™ ready-to-use
 
 
Lipopolysaccharides (from S. abortus equi) S-form, TLRgrade™ ready-to-use
  ENZOALX581150R500
 :  
 
 
Lipopolysaccharides (from S. abortus equi) S-form, TLRgrade™ ready-to-use
  ENZOALX581150R500
 :  

 

  • Pk:
    500 µl
  • Seq:
    1418

 

 

  • Concentration:
    1 mg/ml
  • Formulation:
    Liquid. Sterile, ready-to-use solution in pyrogen-free double-distilled water
  • Purity:
    Absence of detectable protein or DNA contaminants with agonistic TLR activity

 

 

 :
 :GHS08
 :GHS07
 :Refrigerator