Order Entry
Hungary
ContactUsLinkComponent
Lipopolysaccharides (from E.coli) serotype J5 (Rc), TLRgrade™ ready-to-use
Irritant
Health
Lipopolysaccharides (from E.coli) serotype J5 (Rc), TLRgrade™ ready-to-use
Catalog # ENZOALX581014L002
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Irritant
Health
Lipopolysaccharides (from E.coli) serotype J5 (Rc), TLRgrade™ ready-to-use
Catalog # ENZOALX581014L002
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    2 mL
  • Seq:
    2145

Specifications

Specification Test Results

  • Concentration:
    1 mg/ml
  • Formulation:
    Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water
  • Purity:
    Absence of detectable protein or DNA contaminants with agonistic TLR activity

Specification Test Results

About this item

 :
 :GHS07
 :GHS08
Storage Temperature:Refrigerator