Order Entry
Hungary
ContactUsLinkComponent
Lipopolysaccharides (from E.coli) serotype O55:B5, TLRgrade™ ready-to-use
Irritant
Health
Lipopolysaccharides (from E.coli) serotype O55:B5, TLRgrade™ ready-to-use
Supplier:  ENZO LIFE SCIENCES
Certificates
undefined
Irritant
Health
Lipopolysaccharides (from E.coli) serotype O55:B5, TLRgrade™ ready-to-use
Supplier:  ENZO LIFE SCIENCES

Specification Test Results

  • Concentration:
    1 mg/ml
  • Formulation:
    Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water
  • Purity:
    Absence of detectable protein or DNA contaminants with agonistic TLR activity

Specification Test Results

Frequently Bought Together