

Lipopolysaccharides (from E.coli) serotype EH100 (Ra), TLRgrade™ ready-to-use
:
:GHS07
:GHS08
:Refrigerator
- Concentration:1 mg/ml
- Formulation:Liquid. Sterile, ready-to-use solution in double distilled, pyrogen-free water
- Purity:Absence of detectable protein or DNA contaminants with agonistic TLR activity
Frequently Bought Together