Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bufotalin
Bufotalin ≥98% (by HPLC)
Bufotalin ≥98% (by HPLC)
Bufotalin ≥98% (by HPLC)
Catalog # ENZOALX350282M005
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
Bufotalin ≥98% (by HPLC)
Catalog # ENZOALX350282M005
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
CAS Number:
471-95-4
Product Details