0 Ft
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Hungary
ContactUsLinkComponent
Limonin
Limonin ≥98% (by HPLC)
Limonin ≥98% (by HPLC)
Catalog # ENZOALX350225M050
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
Limonin ≥98% (by HPLC)
Catalog # ENZOALX350225M050
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Pk:
50 mg
Seq:
2370
Specifications
About this item
Product Details
:
:
470.52 g/mol
CAS Number:
1180-71-8
Formula:
C₂₆H₃₀O₈
MDL Number:
MFCD00075922
Product Details