Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Anabaenopeptin B
Anabaenopeptin B ≥95% (by HPLC)
Anabaenopeptin B ≥95% (by HPLC)
Anabaenopeptin B ≥95% (by HPLC)
Catalog # ENZOALX350184C100
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
Anabaenopeptin B ≥95% (by HPLC)
Catalog # ENZOALX350184C100
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Pk:
100 µG
Seq:
1286
Specifications
About this item
Product Details
:
:
837.0 g/mol
CAS Number:
161897-74-1
Product Details