Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pyripyropene A
Pyripyropene A ≥85% (by HPLC)
Pyripyropene A ≥85% (by HPLC)
ENZOALX350134C500
:
Enzo Life Sciences
:
Pyripyropene A ≥85% (by HPLC)
ENZOALX350134C500
:
Enzo Life Sciences
:
Pk:
500 µG
Seq:
1418
:
:
583.2 g/mol
:
147444-03-9
:
MFCD00929109