Tentoxin (from Alternaria alternata) ≥98% (by HPLC, TLC)
Catalog # ENZOALX350132MC05
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Kiszerelés:0,5 mg
- Seq:2035
Specifications
About this item
:
:414.5 g/mol
Storage Temperature:Hűtőszekrény
CAS Number:28540-82-1
Formula:C₂₂H₃₀N₄O₄