
Vindoline ≥90% (dry basis)
Catalog # ENZOALX350102M100
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Purity:≥90% (Assay on Dried Basis)
- Source:Isolated from Catharanthus roseus
Specification Test Results
About this item
:
:456.5 g/mol
Merck Index:14,09988
EINECS:218-558-0
:GHS08
Storage Temperature:Freezer
CAS Number:2182-14-1
Formula:C₂₅H₃₂N₂O₆
MDL Number:MFCD27991347