Order Entry
Hungary
ContactUsLinkComponent
 
Ouabain octahydrate ≥98% (by TLC)
Catalog # ENZOALX350066M100
Supplier:  Enzo Life Sciences
CAS Number:  
Ouabain octahydrate ≥98% (by TLC)
Catalog # ENZOALX350066M100
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :584.65 g/mol (anhy)
Storage Temperature:Refrigerator
CAS Number:11018-89-6
 :630-60-4
Formula:C₂₉H₄₄O₁₂.8H₂O
MDL Number:MFCD00149240