
Tetramethylrhodamine ethyl ester perchlorate (TMRE) ≥95% (by HPLC), CELLESTIAL®, Ultrapure mitochondria dye
Catalog # ENZOENZ52309
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:514.95 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:115532-52-0
Formula:C₂₆H₂₇ClN₂O₇
MDL Number:MFCD00467973