(±)4-Hydroxydebrisoquin ≥99%, white solid
Major metabolite of debrisoquin.
Useful indicator for cytochrome P-450 polymorphism. For analytical methods.
Soluble in 1 M HCl.
- :59333-79-8
- :191.2 g/mol
- :Ambient
- Purity:≥99%
Major metabolite of debrisoquin.
Useful indicator for cytochrome P-450 polymorphism. For analytical methods.
Soluble in 1 M HCl.